2-(2-Benzoxazolyl)malondialdehyde 95+%

Supplier: Apollo Scientific

OR7618-25G OR7618-5G OR7618-1G
APOSOR7618-25GEA 845 EUR
APOSOR7618-25G APOSOR7618-5G APOSOR7618-1G
2-(2-Benzoxazolyl)malondialdehyde 95+%
2-(2-Benzoxazolyl)malondialdehyde
Formula: C₁₀H₇NO₃
Melting Pt: 230…232 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00216526
CAS Number: 40071-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR