2-tert-Butyl-1H-indole-3-carbaldehyde 95+%
Supplier: Apollo Scientific
|
Warning
|
OR7623-1G
OR7623-5G
APOSOR7623-1GEA
112
EUR
APOSOR7623-1G
APOSOR7623-5G
2-tert-Butyl-1H-indole-3-carbaldehyde 95+%
2-tert-Butyl-1H-indole-3-carbaldehyde
Formula:
C₁₃H₁₅NO MW: 201,27 g/mol Melting Pt: 224 °C Storage Temperature: Ambient |
MDL Number:
MFCD01241608 CAS Number: 29957-81-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...