2-(3-Chlorophenyl)malonaldehyde 95+%

Supplier: Apollo Scientific

OR7628-25G OR7628-5G OR7628-1G
APOSOR7628-25GEA 1260 EUR
APOSOR7628-25G APOSOR7628-5G APOSOR7628-1G
2-(3-Chlorophenyl)malonaldehyde 95+%
2-(3-Chlorophenyl)malonaldehyde
Formula: C₉H₇ClO₂
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD02261887
CAS Number: 74963-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR