2-(4-Phenylphenyl)malondialdehyde 95+%

Supplier: Apollo Scientific
Warning

OR7653-1G OR7653-5G OR7653-25G
APOSOR7653-1GEA 163 EUR
APOSOR7653-1G APOSOR7653-5G APOSOR7653-25G
2-(4-Phenylphenyl)malondialdehyde 95+%
2-(4-Phenylphenyl)malondialdehyde
Formula: C₁₅H₁₂O₂
Melting Pt: 194…197 °C
Storage Temperature: Ambient
MDL Number: MFCD03426049
CAS Number: 125507-91-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR