1-Acetyl-4-[(2-methoxyethyl)amino]piperidine 95+%
Supplier: Apollo Scientific
|
Warning
|
OR7673-1G
OR7673-5G
OR7673-25G
APOSOR7673-1GEA
68.4
EUR
APOSOR7673-1G
APOSOR7673-5G
APOSOR7673-25G
1-Acetyl-4-[(2-methoxyethyl)amino]piperidine 95+%
1-Acetyl-4-[(2-methoxyethyl)amino]piperidine
Formula:
C₁₀H₂₀N₂O₂ MW: 200,28 g/mol Boiling Pt: 92…97 °C/0,15mmHg Storage Temperature: Ambient |
MDL Number:
MFCD03452783 CAS Number: 849021-41-6 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...