N-[2-(Aminomethyl)phenyl]-N,N-dimethylamine 95+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(Aminomethyl)-N,N-dimethylaniline
OR7676-5G
OR7676-1G
OR7676-250MG
APOSOR7676-5GEA
313
EUR
APOSOR7676-5G
APOSOR7676-1G
APOSOR7676-250MG
N-[2-(Aminomethyl)phenyl]-N,N-dimethylamine 95+%
N-[2-(Aminomethyl)phenyl]-N,N-dimethylamine
Formula:
C₉H₁₄N₂ Boiling Pt: 60 °C/0,037mmHg Storage Temperature: Ambient |
MDL Number:
MFCD04037893 CAS Number: 57678-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...