1-Methyl-3-piperidinemethanol 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-(Hydroxymethyl)-1-methylpiperidine
OR7690-100G
OR7690-25G
OR7690-5G
APOSOR7690-100GEA
106
EUR
APOSOR7690-100G
APOSOR7690-25G
APOSOR7690-5G
1-Methyl-3-piperidinemethanol 98%
1-Methyl-3-piperidinemethanol
Formula:
C₇H₁₅NO MW: 129,2 g/mol Boiling Pt: 140…145 °C Density: 1,013 g/cm³ (25 °C) Flash Pt: 94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006497 CAS Number: 7583-53-1 EINECS: 231-488-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...