2-(Chloromethyl)-1H-imidazole hydrochloride 95+%

Supplier: Apollo Scientific
Warning

OR7713-1G OR7713-5G OR7713-25G
APOSOR7713-1GEA 208 EUR
APOSOR7713-1G APOSOR7713-5G APOSOR7713-25G
2-(Chloromethyl)-1H-imidazole hydrochloride 95+%
2-(Chloromethyl)-1H-imidazole hydrochloride
Formula: C₄H₆Cl₂N₂
Melting Pt: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00233971
CAS Number: 71670-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR