2,3,5-Trichloropyridine 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,3,5-TCP
OR7714-100G
OR7714-500G
APOSOR7714-100GEA
51
EUR
APOSOR7714-100G
APOSOR7714-500G
2,3,5-Trichloropyridine 97%
2,3,5-Trichloropyridine
Formula:
C₅H₂Cl₃N MW: 182,44 g/mol Boiling Pt: 219 °C Melting Pt: 46…50 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043007 CAS Number: 16063-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...