1-(3-Isopropoxyphenyl)methanamine 95+%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Isopropoxybenzylamine

OR7786-1G OR7786-5G OR7786-25G
APOSOR7786-1GEA 235 EUR
APOSOR7786-1G APOSOR7786-5G APOSOR7786-25G
1-(3-Isopropoxyphenyl)methanamine 95+%
1-(3-Isopropoxyphenyl)methanamine
Formula: C₁₀H₁₅NO
MW: 165,24 g/mol
Boiling Pt: 70 °C/0,05mmHg
Storage Temperature: Ambient
MDL Number: MFCD04037891
CAS Number: 400771-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR