Methyl-4-(3-methoxy-3-oxopropanoyl)benzoate 95+%

Supplier: Apollo Scientific
Danger

Synonyms: Methyl 4-methoxycarbonylbenzoylacetate

OR7815-1G OR7815-5G OR7815-25G
APOSOR7815-1GEA 143 EUR
APOSOR7815-1G APOSOR7815-5G APOSOR7815-25G
Methyl-4-(3-methoxy-3-oxopropanoyl)benzoate 95+%
Methyl-4-(3-methoxy-3-oxopropanoyl)benzoate
Formula: C₁₂H₁₂O₅
MW: 236,223 g/mol
Melting Pt: 81…86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00800637
CAS Number: 22027-52-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR