3-[Acetyl(methyl)amino]propanoic acid 95+%

Supplier: Apollo Scientific
Warning

OR7817-1G OR7817-500MG
APOSOR7817-1GEA 0 EUR
APOSOR7817-1G APOSOR7817-500MG
3-[Acetyl(methyl)amino]propanoic acid 95+%
3-[Acetyl(methyl)amino]propanoic acid
Formula: C₆H₁₁NO₃
Melting Pt: 91…92 °C
Storage Temperature: Ambient
MDL Number: MFCD04037898
CAS Number: 58706-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR