1-(5-Bromo-2-pyridinyl)-4-methylpiperazine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-(5-Bromopyridin-2-yl)-4-methylpiperazine, 5-Bromo-2-(4-methylpiperazin-1-yl)pyridine
OR7840-1G
OR7840-5G
APOSOR7840-1GEA
95.7
EUR
APOSOR7840-1G
APOSOR7840-5G
1-(5-Bromo-2-pyridinyl)-4-methylpiperazine
1-(5-Bromo-2-pyridinyl)-4-methylpiperazine
Formula:
C₁₀H₁₄BrN₃ MW: 256,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07368225 CAS Number: 364794-58-1 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...