5-Bromo-2-(methylamino)pyrimidine

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromo-N-methylpyrimidin-2-amine

OR7851-1G OR7851-5G
APOSOR7851-1GEA 53.9 EUR
APOSOR7851-1G APOSOR7851-5G
5-Bromo-2-(methylamino)pyrimidine
5-Bromo-2-(methylamino)pyrimidine
Formula: C₅H₆BrN₃
MW: 188,0300 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368227
CAS Number: 31402-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR