1,3,5-Triphenylbenzene

Supplier: Apollo Scientific
Warning

OR7856-250G OR7856-50G OR7856-100G
APOSOR7856-250GEA 125 EUR
APOSOR7856-250G APOSOR7856-50G APOSOR7856-100G
1,3,5-Triphenylbenzene
1,3,5-Triphenylbenzene
Formula: C₂₄H₁₈
MW: 306,41 g/mol
Boiling Pt: 460 °C
Melting Pt: 172…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00003060
CAS Number: 612-71-5
EINECS: 210-318-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR