4-Nitroindole 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Nitro-1H-indole
OR7858-1G
OR7858-5G
OR7858-25G
APOSOR7858-1GEA
21.9
EUR
APOSOR7858-1G
APOSOR7858-5G
APOSOR7858-25G
4-Nitroindole 97%
4-Nitroindole
Formula:
C₈H₆N₂O₂ MW: 162,15 g/mol Melting Pt: 202…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010056 CAS Number: 4769-97-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...