N-Benzylanthranilic acid
Supplier: Apollo Scientific
|
Warning
|
OR7874-25G
OR7874-50G
APOSOR7874-25GEA
155
EUR
APOSOR7874-25G
APOSOR7874-50G
N-Benzylanthranilic acid
N-Benzylanthranilic acid
Formula:
C₁₄H₁₃NO₂ Melting Pt: 172…177 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020247 CAS Number: 6622-55-5 EINECS: 229-572-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...