8-Bromoquinoline 98%

Supplier: Apollo Scientific
Warning

OR7885-25G OR7885-5G
APOSOR7885-25GEA 65.4 EUR
APOSOR7885-25G APOSOR7885-5G
8-Bromoquinoline 98%
8-Bromoquinoline
Formula: C₉H₆BrN
MW: 208,06 g/mol
Boiling Pt: 112…113 °C/0,5mmHg
Density: 1,594 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00191859
CAS Number: 16567-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR