3'-Nitroformanilide 98%
Supplier: Apollo Scientific
OR7891-100G
OR7891-25G
APOSOR7891-100GEA
216
EUR
APOSOR7891-100G
APOSOR7891-25G
3'-Nitroformanilide 98%
3'-Nitroformanilide
Formula:
C₇H₆N₂O₃ Melting Pt: 132…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017014 CAS Number: 102-38-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...