Methyl-1H-indene-3-carboxylate 95+%
Supplier: Apollo Scientific
|
Warning
|
OR7915-1G
OR7915-250MG
APOSOR7915-1GEA
538
EUR
APOSOR7915-1G
APOSOR7915-250MG
Methyl-1H-indene-3-carboxylate 95+%
Methyl-1H-indene-3-carboxylate
Formula:
C₁₁H₁₀O₂ MW: 174.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04037885 CAS Number: 39891-79-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...