3,5-Bis(methylsulphonyl)benzamide 95+%
Supplier: Apollo Scientific
OR7951-1G
OR7951-5G
OR7951-25G
APOSOR7951-1GEA
229
EUR
APOSOR7951-1G
APOSOR7951-5G
APOSOR7951-25G
3,5-Bis(methylsulphonyl)benzamide 95+%
3,5-Bis(methylsulphonyl)benzamide
Formula:
C₉H₁₁NO₅S₂ MW: 277.32 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04037978 CAS Number: 849924-85-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...