4-Bromo-2-(methylsulphonyl)benzaldehyde

Supplier: Apollo Scientific
Warning

OR7961-10G OR7961-1G OR7961-5G
APOSOR7961-10GEA 481 EUR
APOSOR7961-10G APOSOR7961-1G APOSOR7961-5G
4-Bromo-2-(methylsulphonyl)benzaldehyde
4-Bromo-2-(methylsulphonyl)benzaldehyde
Formula: C₈H₇BrO₃S
MW: 263,11 g/mol
Melting Pt: 108…110 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD04037947
CAS Number: 849035-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR