1-(2-Pyrimidyl)piperazine 99%

Supplier: Apollo Scientific
Warning

Synonyms: 2-(1-Piperazinyl)pyrimidine, 2-(Piperazin-1-yl)pyrimidine

OR8006-100G OR8006-25G
APOSOR8006-100GEA 87 EUR
APOSOR8006-100G APOSOR8006-25G
1-(2-Pyrimidyl)piperazine 99%
1-(2-Pyrimidyl)piperazine
Formula: C₈H₁₂N₄
MW: 164,21 g/mol
Boiling Pt: 277 °C
Melting Pt: 31…35 °C
Density: 1,158 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00040742
CAS Number: 20980-22-7
EINECS: 244-135-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR