1,4-Benzodioxane-2-carbonyl chloride 97%

Supplier: Apollo Scientific
Danger

OR8020-1G
APOSOR8020-1GEA 184 EUR
APOSOR8020-1G
1,4-Benzodioxane-2-carbonyl chloride 97%
1,4-Benzodioxane-2-carbonyl chloride
Formula: C₉H₇ClO₃
Melting Pt: 56…59 °C
Storage Temperature: Ambient
MDL Number: MFCD02180342
CAS Number: 3663-81-8
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR