4-Bromo-1-butene 99%
Supplier: Apollo Scientific
|
Danger
|
OR8033-100G
OR8033-25G
OR8033-500G
APOSOR8033-100GEA
62.6
EUR
APOSOR8033-100G
APOSOR8033-25G
APOSOR8033-500G
4-Bromo-1-butene 99%
4-Bromo-1-butene
Formula:
C₄H₇Br MW: 135 g/mol Boiling Pt: 98…100 °C Density: 1,33 g/cm³ (25 °C) Flash Pt: 1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000258 CAS Number: 5162-44-7 EINECS: 225-937-4 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...