Ethyl-2-amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate 97%

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 2-amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate

OR8091-1G OR8091-250MG
APOSOR8091-1GEA 132 EUR
APOSOR8091-1G APOSOR8091-250MG
Ethyl-2-amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate 97%
Ethyl-2-amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate
Formula: C₁₁H₁₆N₂O₂S
MW: 240,33 g/mol
Melting Pt: 100…102 °C
Storage Temperature: Ambient
MDL Number: MFCD01147322
CAS Number: 24237-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR