6-Bromoimidazo[1,2-a]pyrimidine
Supplier: Apollo Scientific
|
Warning
|
OR8122-1G
OR8122-250MG
OR8122-5G
APOSOR8122-1GEA
31.9
EUR
APOSOR8122-1G
APOSOR8122-250MG
APOSOR8122-5G
6-Bromoimidazo[1,2-a]pyrimidine
6-Bromoimidazo[1,2-a]pyrimidine
Formula:
C₆H₄BrN₃ MW: 198,0200 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09261434 CAS Number: 865156-68-9 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...