3,6-Dichloropicolinic acid
Supplier: Apollo Scientific
|
Danger
|
OR8123-100G
OR8123-25G
OR8123-500G
APOSOR8123-100GEA
58
EUR
APOSOR8123-100G
APOSOR8123-25G
APOSOR8123-500G
3,6-Dichloropicolinic acid
3,6-Dichloropicolinic acid
Formula:
C₆H₃Cl₂NO₂ Melting Pt: 150 °C (dec.) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00078655 CAS Number: 1702-17-6 EINECS: 216-935-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...