1-Methylimidazole-4-sulphonamide 97%
Supplier: Apollo Scientific
|
Warning
|
OR8127-1G
OR8127-250MG
OR8127-5G
APOSOR8127-1GEA
147
EUR
APOSOR8127-1G
APOSOR8127-250MG
APOSOR8127-5G
1-Methylimidazole-4-sulphonamide 97%
1-Methylimidazole-4-sulphonamide
Formula:
C₄H₇N₃O₂S Melting Pt: 222…224 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02091382 CAS Number: 111124-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...