6-Amino-m-cresol 98%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Amino-2-hydroxy-4-methylbenzene, 2-Hydroxy-4-methylaniline, Cetilistat Impurity 4, 4-Amino-3-hydroxytoluene, 2-Amino-5-methylphenol

OR8233-100G OR8233-25G
APOSOR8233-100GEA 158 EUR
APOSOR8233-100G APOSOR8233-25G
6-Amino-m-cresol 98%
6-Amino-m-cresol
Formula: C₇H₉NO
MW: 123,15 g/mol
Melting Pt: 159…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00007693
CAS Number: 2835-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR