4-Chloro-7-methoxyquinoline
Supplier: Apollo Scientific
|
Warning
|
OR8277-1G
OR8277-250MG
APOSOR8277-1GEA
43.7
EUR
APOSOR8277-1G
APOSOR8277-250MG
4-Chloro-7-methoxyquinoline
4-Chloro-7-methoxyquinoline
Formula:
C₁₀H₈ClNO MW: 193,6300 g/mol Melting Pt: 75…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659036 CAS Number: 68500-37-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...