4-Methyl-2-thiopheneboronic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: (4-Methylthiophen-2-yl)boronic acid, 4-Methylthiophene-2-boronic acid

OR8282-25G OR8282-5G OR8282-100MG OR8282-10G
APOSOR8282-25GEA 590 EUR
APOSOR8282-25G APOSOR8282-5G APOSOR8282-100MG APOSOR8282-10G
4-Methyl-2-thiopheneboronic acid 97%
4-Methyl-2-thiopheneboronic acid
Formula: C₅H₇BO₂S
MW: 141,99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01319040
CAS Number: 162607-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR