6-Chloro-4-quinazolone 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Chloroquinazolin-4-ol
OR8300-1G
OR8300-5G
APOSOR8300-1GEA
69.8
EUR
APOSOR8300-1G
APOSOR8300-5G
6-Chloro-4-quinazolone 98%
6-Chloro-4-quinazolone
Formula:
C₈H₅ClN₂O MW: 180,5900 g/mol Melting Pt: 271…274 °C Storage Temperature: Ambient |
MDL Number:
MFCD01686365 CAS Number: 16064-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...