N-Methylcyclohexylamine
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
N-Cyclohexylmethylamine, Cyclohexyl(methyl)amine
OR8357-100G
OR8357-5G
APOSOR8357-100GEA
43.7
EUR
APOSOR8357-100G
APOSOR8357-5G
N-Methylcyclohexylamine
N-Methylcyclohexylamine
Formula:
C₇H₁₅N MW: 113,2 g/mol Boiling Pt: 149 °C Melting Pt: –9…–7 °C Density: 0,868 g/cm³ (25 °C) Flash Pt: 29 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003832 CAS Number: 100-60-7 EINECS: 202-869-3 UN: 2734 ADR: 8+3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...