2-Phenylpyrimidine
Supplier: Apollo Scientific
|
Warning
|
OR8422-1G
APOSOR8422-1GEA
258
EUR
APOSOR8422-1G
2-Phenylpyrimidine
2-Phenylpyrimidine
Formula:
C₁₀H₈N₂ MW: 156,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04038761 CAS Number: 7431-45-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...