3,4,5-Trimethoxybenzyl chloride 95%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
3,4,5-Trimethoxybenzylchloride, 5-(Chloromethyl)-1,2,3-trimethoxybenzene
OR8473-5G
OR8473-1G
APOSOR8473-5GEA
53.9
EUR
APOSOR8473-5G
APOSOR8473-1G
3,4,5-Trimethoxybenzyl chloride 95%
3,4,5-Trimethoxybenzyl chloride
Formula:
C₁₀H₁₃ClO₃ MW: 216,66 g/mol Melting Pt: 60…63 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00040716 CAS Number: 3840-30-0 EINECS: 223-330-9 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...