(1-Benzyl-1H-1,2,3-triazol-4-yl)methanol

Supplier: Apollo Scientific
Warning

OR8490-1G OR8490-500MG OR8490-5G
APOSOR8490-1GEA 222 EUR
APOSOR8490-1G APOSOR8490-500MG APOSOR8490-5G
(1-Benzyl-1H-1,2,3-triazol-4-yl)methanol
(1-Benzyl-1H-1,2,3-triazol-4-yl)methanol
Formula: C₁₀H₁₁N₃O
Melting Pt: 74…76 °C
Storage Temperature: Ambient
MDL Number: MFCD09607905
CAS Number: 28798-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR