5-(2-Bromophenoxy)-2-furoic acid

Supplier: Apollo Scientific

OR8493-1G OR8493-5G
APOSOR8493-1GEA 336 EUR
APOSOR8493-1G APOSOR8493-5G
5-(2-Bromophenoxy)-2-furoic acid
5-(2-Bromophenoxy)-2-furoic acid
Formula: C₁₁H₇BrO₄
Melting Pt: 145…147 °C
Storage Temperature: Ambient
MDL Number: MFCD09607936
CAS Number: 952183-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR