Ethyl-5-amino-1-benzofuran-2-carboxylate 99%
|
Warning
|
OR8508-25G
OR8508-5G
OR8508-10G
APOSOR8508-25GEA
285
EUR
APOSOR8508-25G
APOSOR8508-5G
APOSOR8508-10G
Ethyl-5-amino-1-benzofuran-2-carboxylate 99%
Ethyl-5-amino-1-benzofuran-2-carboxylate
Formula:
C₁₁H₁₁NO₃ MW: 205,22 g/mol Melting Pt: 54…65 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08275091 CAS Number: 174775-48-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...