1-Boc-4-(5-bromopyrimidin-2-yl)piperazine

Supplier: Apollo Scientific
Warning

Synonyms: tert-Butyl 4-(5-bromopyrimidin-2-yl)piperazine-1-carboxylate

OR8545-1G OR8545-5G
APOSOR8545-1GEA 50.9 EUR
APOSOR8545-1G APOSOR8545-5G
1-Boc-4-(5-bromopyrimidin-2-yl)piperazine
1-Boc-4-(5-bromopyrimidin-2-yl)piperazine
Formula: C₁₃H₁₉BrN₄O₂
MW: 343,23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07375175
CAS Number: 374930-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR