4-Boc-1-(5-bromo-2-pyridyl)piperazine

OR8546-1G OR8546-5G OR8546-25G
APOSOR8546-1GEA 65.5 EUR
APOSOR8546-1G APOSOR8546-5G APOSOR8546-25G
4-Boc-1-(5-bromo-2-pyridyl)piperazine
4-Boc-1-(5-bromo-2-pyridyl)piperazine
Formula: C₁₄H₂₀BrN₃O₂
MW: 342,237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07369772
CAS Number: 153747-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR