6-Methoxyisoquinoline

Supplier: Apollo Scientific
Warning

OR8562-1G OR8562-5G OR8562-10G
APOSOR8562-1GEA 21.9 EUR
APOSOR8562-1G APOSOR8562-5G APOSOR8562-10G
6-Methoxyisoquinoline
6-Methoxyisoquinoline
Formula: C₁₀H₉NO
Boiling Pt: 88…95 °C/0,8mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD04114861
CAS Number: 52986-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR