8-Methoxyisoquinoline 98%

Supplier: Apollo Scientific
Warning

OR8564-1G OR8564-250MG OR8564-5G
APOSOR8564-1GEA 326 EUR
APOSOR8564-1G APOSOR8564-250MG APOSOR8564-5G
8-Methoxyisoquinoline 98%
8-Methoxyisoquinoline
Formula: C₁₀H₉NO
Melting Pt: 48…52 °C
Storage Temperature: Ambient
MDL Number: MFCD09258766
CAS Number: 1723-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR