1-(4-Bromo-phenyl)-1H-pyrazole 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-(4-Bromophenyl)-1H-pyrazole
OR8582-1G
OR8582-5G
APOSOR8582-1GEA
58.2
EUR
APOSOR8582-1G
APOSOR8582-5G
1-(4-Bromo-phenyl)-1H-pyrazole 98%
1-(4-Bromo-phenyl)-1H-pyrazole
Formula:
C₉H₇BrN₂ MW: 223,08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08059318 CAS Number: 13788-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...