4-Bromo-2-(trimethylsilyl)thiazole 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Bromo-2-trimethylsilanylthiazole
OR8607-1G
OR8607-5G
APOSOR8607-1GEA
162
EUR
APOSOR8607-1G
APOSOR8607-5G
4-Bromo-2-(trimethylsilyl)thiazole 95%
4-Bromo-2-(trimethylsilyl)thiazole
Formula:
C₆H₁₀BrNSSi MW: 236,21 g/mol Density: 1,384 g/cm³ (25 °C) Flash Pt: 82,2 °C Storage Temperature: Ambient |
MDL Number:
MFCD01319107 CAS Number: 108306-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...