Methyl-4-methoxysalicylate 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 2-hydroxy-4-methoxybenzoate
OR8647-100G
OR8647-25G
OR8647-500G
APOSOR8647-100GEA
219
EUR
APOSOR8647-100G
APOSOR8647-25G
APOSOR8647-500G
Methyl-4-methoxysalicylate 98%
Methyl 4-methoxysalicylate
Formula:
C₉H₁₀O₄ MW: 182,18 g/mol Boiling Pt: 134 °C/ 7mmHg Melting Pt: 47…51 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008424 CAS Number: 5446-02-6 EINECS: 226-657-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...