2-(2-Dimethylaminoethoxy)benzoic acid 95%
Supplier: Apollo Scientific
|
Warning
|
OR8666-1G
OR8666-5G
APOSOR8666-1GEA
207
EUR
APOSOR8666-1G
APOSOR8666-5G
2-(2-Dimethylaminoethoxy)benzoic acid 95%
2-(2-Dimethylaminoethoxy)benzoic acid
Formula:
C₁₁H₁₅NO₃ MW: 209,25 g/mol Melting Pt: 108…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD07801180 CAS Number: 206261-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...