Furan-2-boronic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: Furan-2-ylboronic acid

OR8669-100G OR8669-25G OR8669-5G
APOSOR8669-100GEA 174 EUR
APOSOR8669-100G APOSOR8669-25G APOSOR8669-5G
Furan-2-boronic acid 98%
Furan-2-boronic acid
Formula: C₄H₅BO₃
MW: 111.89 g/mol
Melting Pt: 120 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00799544
CAS Number: 13331-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR