3-(Pyrrolidin-1-ylmethyl)benzaldehyde 97%
Supplier: Apollo Scientific
|
Danger
|
OR8677-1G
OR8677-250MG
APOSOR8677-1GEA
317
EUR
APOSOR8677-1G
APOSOR8677-250MG
3-(Pyrrolidin-1-ylmethyl)benzaldehyde 97%
3-(Pyrrolidin-1-ylmethyl)benzaldehyde
Formula:
C₁₂H₁₅NO MW: 189,26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07782420 CAS Number: 884507-42-0 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...