3-Bromo-2-iodopyridine
Supplier: Apollo Scientific
|
Warning
|
OR8711-1G
OR8711-5G
APOSOR8711-1GEA
109
EUR
APOSOR8711-1G
APOSOR8711-5G
3-Bromo-2-iodopyridine
3-Bromo-2-iodopyridine
Formula:
C₅H₃BrIN Boiling Pt: 138…139 °C/8mmHg Melting Pt: 50…52 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08543471 CAS Number: 408502-43-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...